3,5-Dichlorthiophenol 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3,5-Dichlorphenylmercaptan
435680010
435680050
ACRO435680010EA
32.1
EUR
ACRO435680010
ACRO435680050
3,5-Dichlorthiophenol 97%
3,5-Dichlorthiophenol
Formel:
C₆H₄Cl₂S Molecular Weight: 179.07 g/mol Siedepunkt: 125…130 °C (12 mmHg) Schmelzpunkt: 62…65 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041423 CAS-Nummer: 17231-94-6 UN: 3335 ADR: 9 III |
Spezifikation Testergebnisse
Appearance | White to yellow Crystalline powder or solid |
Infrared spectrum | Conforms |
Melting point | 62 to 65 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...