Diisopropylaminomethylidene(diisopropyl)ammonium tetrafluoroborate 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Di-i-propylaminomethylene(di-i-propyl)aminium tetrafluoroborate, 2-(Diisopropylamino)-1,1-diisopropyl-1-azoniaethene, N-((Diisopropylamino)methylene)-N-isopropylpropan-2-aminium tetrafluoroborate, N-((DIISOPROPYLAMINO)METHYLENE)-N-DIISOPROPYLAMINIUM TETRAFLUOROBORATE
435700010
ACRO435700010EA
53.1
EUR
ACRO435700010
Diisopropylaminomethylidene(diisopropyl)ammonium tetrafluoroborate 97%
Diisopropylaminomethylidene(diisopropyl)ammonium tetrafluoroborate
Formel:
C₁₃H₂₉BF₄N₂ Molecular Weight: 300,19 g/mol Schmelzpunkt: 280…284 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD15144860 CAS-Nummer: 369405-27-6 |
Spezifikation Testergebnisse
Appearance | White to beige Powder or solid |
Infrared spectrum | Conforms |
NMR | ≥96 % |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...