3-Amino-4-iod-1H-indazol 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Iodo-1H-indazol-3-amine
435750050
ACRO435750050EA
124
EUR
ACRO435750050
3-Amino-4-iod-1H-indazol 95%
3-Amino-4-iodo-1H-indazole
Formel:
C₇H₆IN₃ Molecular Weight: 259,0471 g/mol Schmelzpunkt: 150…152 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03426697 CAS-Nummer: 599191-73-8 |
Spezifikation Testergebnisse
Appearance | Off-white to brown Crystalline solid |
Infrared spectrum | Conforms |
Melting point | 150 to 155 °C |
HPLC | ≥94.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...