3-Amino-6-brom-1H-indazol 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 6-Bromo-1H-indazol-3-amine

435780050
ACRO435780050EA 207 EUR
ACRO435780050
3-Amino-6-brom-1H-indazol 95%
3-Amino-6-bromo-1H-indazole
Formel: C₇H₆BrN₃
Molecular Weight: 212,05 g/mol
Schmelzpunkt: 175…178 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05665872
CAS-Nummer: 404827-77-6
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 235 to 240 °C
HPLC ≥94.0 %
Specific optical rotation +40° to +44° (20 °C, 589 nm) (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR