(R)-Tetrahydrofuran-3-amine-4-methylbenzenesulphonate 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (R)-(+)-3-Aminotetrahydrofuran p-toluenesulphonate salt, (R)-(-)-3-aminotetrahydrofurantoluene-4-sulphonate, (R)-(+)-3-Aminotetrahydrofuran p-toluenesulphonate, (R)-Tetrahydrofuran-3-amine 4-methylbenzenesulphonate, (R)-Tetrahydrofuran-3-amine-4-methylbenzenesulphonate

435820050
ACRO435820050EA 500 EUR
ACRO435820050
(R)-Tetrahydrofuran-3-amine-4-methylbenzenesulphonate 95%
(R)-Tetrahydrofuran-3-amine-4-methylbenzenesulphonate
Formel: C₁₁H₁₇NO₄S
Molecular Weight: 259.32 g/mol
Schmelzpunkt: 137.3…141 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00040622
CAS-Nummer: 111769-27-8

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 136 to 143 °C
Elemental Analysis 5.18 to 5.62 % N
Specific optical rotation +3° to +5° (20 °C, 589 nm) (c=1, methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR