Isoindoline hydrochloride 97%

Lieferant: Thermo Fisher Scientific
Achtung

435850050 435850250
ACRO435850050EA 98 EUR
ACRO435850050 ACRO435850250
Isoindoline hydrochloride 97%
Isoindoline hydrochloride
Formel: C₈H₁₀ClN
Molecular Weight: 155.625 g/mol
Schmelzpunkt: 255…256 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD03502377
CAS-Nummer: 32372-82-0

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white to grey Powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Elemental Analysis 8.64 to 9.36 % N


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR