2-Brom-4-pyridinboronsäure 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Brompyridin-4-boronsäure, 2-Brom-4-pyridinylboronsäure

435920010
ACRO435920010EA 524 EUR
ACRO435920010
2-Brom-4-pyridinboronsäure 95%
2-Bromo-4-pyridineboronic acid
Formel: C₅H₅BBrNO₂
Molecular Weight: 201,81 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06798244
CAS-Nummer: 458532-94-0

Bestellen

Spezifikation Testergebnisse

Appearance White to brown Crystalline powder (May have a pink hue)
Infrared spectrum Conforms
HPLC ≥94.0 %
GC ≥96.0 %
Specific optical rotation +16° to +20° (20°C, 589 nm) (c=5% in Methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR