Nortropinone hydrochloride 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 8-Azabicyclo[3.2.1]octan-3-one hydrochloride

436120010 436120050
ACRO436120010EA 79.4 EUR
ACRO436120010 ACRO436120050
Nortropinone hydrochloride 97%
Nortropinone hydrochloride
Formel: C₇H₁₂ClNO
Schmelzpunkt: 194,7…195.8 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD03613582
CAS-Nummer: 25602-68-0

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white to light brown Powder or solid
Infrared spectrum Conforms
Titration Argentometric ≥96.0 to ≤104.0 %
GC ≥98.0 %
Specific optical rotation +12° to +15° (20°C, 589 nm) (Neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR