(R)-(-)-Methyl-3-hydroxyisobutyrat 98%

Lieferant: Thermo Scientific

Synonyme: Methyl (R)-(-)-beta-hydroxyisobutyrate, (R)-Methyl 3-hydroxy-2-methylpropanoate

436180050
ACRO436180050EA 144 EUR
ACRO436180050
(R)-(-)-Methyl-3-hydroxyisobutyrat 98%
(R)-(-)-Methyl-3-hydroxyisobutyrat
Formel: C₅H₁₀O₃
Molecular Weight: 118,1311 g/mol
Siedepunkt: 60 °C (4Torr)
Dichte: 1,07 g/cm³
Flammpunkt: 81 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00063450
CAS-Nummer: 72657-23-9

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥97.5 %
Specific optical rotation -23° to -29° (20°C, 589 nm) (c=4, methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR