Acetobromo-α-D-glucuronic acid methyl ester 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
BROMOMETHYLTRIACETYL GLUCURONATE, Acetobromo-α-D-glucuronic Acid Methyl Ester, Methyl acetobromo-α-D-glucuronate, (2,3,4-Tri-O-acetyl-α-D-glucopyranosyl bromide)uronic acid methyl ester, (2R,3R,4S,5S,6S)-2-Bromo-6-(methoxycarbonyl)tetrahydro-2H-pyran-3,4,5-triyl triacetate
436290010
436290050
ACRO436290010EA
190
EUR
ACRO436290010
ACRO436290050
Acetobromo-α-D-glucuronic acid methyl ester 98%
Acetobromo-alpha-D-glucuronic acid methyl ester
Formel:
C₁₃H₁₇BrO₉ Molecular Weight: 397.174 g/mol Schmelzpunkt: 80…110 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00061613 CAS-Nummer: 21085-72-3 |
Spezifikation Testergebnisse
Appearance | White to grey Crystalline powder or solid |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % (ELSD) |
Stabilizer | 2 % CaCO3 |
Specific optical rotation | +185° to +195° (20°C, 589 nm) (c=1, ethyl acetate) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...