3-Fluor-4-iodpyridin 97%

Lieferant: Thermo Fisher Scientific
Achtung

436380050
ACRO436380050EA 351 EUR
ACRO436380050
3-Fluor-4-iodpyridin 97%
3-Fluor-4-iodpyridin
Formel: C₅H₃FIN
Molecular Weight: 222.99 g/mol
Schmelzpunkt: 189.4…190.4 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03092927
CAS-Nummer: 22282-75-3

Bestellen

Spezifikation Testergebnisse

Appearance Cream to beige Crystalline powder
Infrared spectrum Conforms
Melting point 87 to 91 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR