(R)-3-(Aminomethyl)-1-Boc-piperidin 97%

Lieferant: Thermo Fisher Scientific
Achtung

436420010 436420050
ACRO436420010EA 94.9 EUR
ACRO436420010 ACRO436420050
(R)-3-(Aminomethyl)-1-Boc-piperidin 97%
(R)-3-(Aminomethyl)-1-Boc-piperidin
Formel: C₁₁H₂₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03839876
CAS-Nummer: 140645-23-4

Bestellen

Spezifikation Testergebnisse

Appearance Colorless to light yellow Liquid or low melting solid
Infrared spectrum Conforms
GC ≥96.0 %
Specific optical rotation +10° to +20° (20°C, 589 nm) (c=1 in MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR