(R)-3-(Aminomethyl)-1-Boc-piperidin 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
436420010
436420050
ACRO436420010EA
94.9
EUR
ACRO436420010
ACRO436420050
(R)-3-(Aminomethyl)-1-Boc-piperidin 97%
(R)-3-(Aminomethyl)-1-Boc-piperidin
Formel:
C₁₁H₂₂N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD03839876 CAS-Nummer: 140645-23-4 |
Spezifikation Testergebnisse
Appearance | Colorless to light yellow Liquid or low melting solid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Specific optical rotation | +10° to +20° (20°C, 589 nm) (c=1 in MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...