(R)-1-Boc-3-hydroxypiperidin 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
436430050
ACRO436430050EA
276
EUR
ACRO436430050
(R)-1-Boc-3-hydroxypiperidin 97%
(R)-1-Boc-3-hydroxypiperidin
Formel:
C₁₀H₁₉NO₃ Schmelzpunkt: 40…42 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115306 CAS-Nummer: 143900-43-0 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | White to tan Low melting solid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Specific optical rotation | -8.5° to -12.0° (20°C, 589 nm) (c=1 in CHCl3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...