(S)-1-Boc-3-hydroxypiperidin 97%

Lieferant: Thermo Fisher Scientific
Achtung

436440050
ACRO436440050EA 270 EUR
ACRO436440050
(S)-1-Boc-3-hydroxypiperidin 97%
(S)-1-Boc-3-hydroxypiperidin
Formel: C₁₀H₁₉NO₃
Flammpunkt: 110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115307
CAS-Nummer: 143900-44-1

Bestellen

Spezifikation Testergebnisse

Appearance White to tan Powder
Infrared spectrum Conforms
GC ≥96.0 %
Specific optical rotation +9° to +11° (20°C, 589 nm) (C=1 in chloroform) / +21° to +24° (20°C, 589 nm) (c=5 in MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR