(R)-(-)-3-Amino-1-Boc-piperidin 97%

Lieferant: Thermo Fisher Scientific
Achtung

436490050
ACRO436490050EA 531 EUR
ACRO436490050
(R)-(-)-3-Amino-1-Boc-piperidin 97%
(R)-(-)-3-Amino-1-Boc-piperidin
Formel: C₁₀H₂₀N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03094717
CAS-Nummer: 188111-79-7

Bestellen

Spezifikation Testergebnisse

Appearance Colorless to light yellow Liquid or low melting solid
Infrared spectrum Conforms
GC ≥96.0 %
Specific optical rotation -35° to -38° (20°C, 589 nm) (c=5 in MeOH) / -26.5° to -30.5° (20°C, 589 nm) (c=1, DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR