(R)-(-)-tert-Leucinol 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: D-tert-Leucinol, (2S)-2-Amino-3,3-dimethyl-1-hydroxybutane, (R)-2-Amino-3,3-dimethylbutan-1-ol

436671000
ACRO436671000EA 111 EUR
ACRO436671000
(R)-(-)-tert-Leucinol 98%
(R)-(-)-tert-Leucinol
Formel: C₆H₁₅NO
Molecular Weight: 117,19 g/mol
Siedepunkt: 70 °C (,5hPa)
Dichte: 0,9 g/cm³
Flammpunkt: 90 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01076574
CAS-Nummer: 112245-09-7

Bestellen

Spezifikation Testergebnisse

Appearance Colorless to yellow Liquid or low melting solid
Infrared spectrum Conforms
GC ≥97.5 %
Specific optical rotation -34° to -40° (20°C, 589 nm) (c=0.7, ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR