Naproxen sodium 98%

Lieferant: Thermo Fisher Scientific
Gefahr

436710050 436710250
ACRO436710050EA 81.1 EUR
ACRO436710050 ACRO436710250
Naproxen sodium 98%
Naproxen sodium
Formel: C₁₄H₁₃NaO₃
Schmelzpunkt: 259…262 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00058507
CAS-Nummer: 26159-34-2

Bestellen

Spezifikation Testergebnisse

Appearance Colorless to light yellow Powder
Infrared spectrum Conforms
Specific optical rotation -14.7° to -17.0° (20°C, 589 nm) (c=5, 1N NaOH)
Titration with HClO4 98.0 to 102.0 %
Free acid ≤1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR