Eisen 99.99% Trace metals basis, granules
Lieferant: Thermo Fisher Scientific
437110050
437110250
ACRO437110050EA
47.4
EUR
ACRO437110050
ACRO437110250
Eisen 99.99% Trace metals basis, granules
Iron
Formel:
Fe Molecular Weight: 55.85 g/mol Siedepunkt: 3000 °C (760 mmHg) Schmelzpunkt: 1535 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010999 CAS-Nummer: 7439-89-6 Merck Index: 13,05109 |
Spezifikation Testergebnisse
Appearance | Silver grey Granules |
Particle size | 0.5 to 1 mm |
Total trace metal impurities | ≤100 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...