Isoproterenol hydrochloride 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: DL-Isoproterenol hydrochloride, (±)-Isoproterenol hydrochloride, Isoprenaline hydrochloride

437210050 437210250 437211000
ACRO437210050EA 47.7 EUR
ACRO437210050 ACRO437210250 ACRO437211000
Isoproterenol hydrochloride 98%
Isoproterenol hydrochloride
Formel: C₁₁H₁₇NO₃
Molecular Weight: 247.72 g/mol
Schmelzpunkt: 165…175 °C (Zers.)
Lagertemperatur: Kühlschrank
MDL: MFCD00012603
CAS-Nummer: 51-30-9

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 164 to 170 °C
Loss on drying ≤1 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR