Isoproterenol hydrochloride 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
DL-Isoproterenol hydrochloride, (±)-Isoproterenol hydrochloride, Isoprenaline hydrochloride
437210050
437210250
437211000
ACRO437210050EA
47.7
EUR
ACRO437210050
ACRO437210250
ACRO437211000
Isoproterenol hydrochloride 98%
Isoproterenol hydrochloride
Formel:
C₁₁H₁₇NO₃ Molecular Weight: 247.72 g/mol Schmelzpunkt: 165…175 °C (Zers.) Lagertemperatur: Kühlschrank |
MDL:
MFCD00012603 CAS-Nummer: 51-30-9 |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | 164 to 170 °C |
Loss on drying | ≤1 % |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...