2-Bromisonicotinsäure 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Bromo-4-pyridinecarboxylic acid, 2-Bromopyridine-4-carboxylic acid
437380050
ACRO437380050EA
162
EUR
ACRO437380050
2-Bromisonicotinsäure 97%
2-Bromisonicotinsäure
Formel:
C₆H₄BrNO₂ Molecular Weight: 202.01 g/mol Schmelzpunkt: 212…222 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01646069 CAS-Nummer: 66572-56-3 |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...