5-Bromo-2-methoxypyridine-3-carboxaldehyde 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Bromo-2-methoxynicotinaldehyde
437390050
ACRO437390050EA
510
EUR
ACRO437390050
5-Bromo-2-methoxypyridine-3-carboxaldehyde 97%
5-Bromo-2-methoxypyridine-3-carboxaldehyde
Formel:
C₇H₆BrNO₂ Molecular Weight: 216,034 g/mol Schmelzpunkt: 102…104 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05663521 CAS-Nummer: 103058-87-3 |
Spezifikation Testergebnisse
Appearance | White to brown Powder |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...