(S)-3-(Aminomethyl)-1-Boc-piperidin 97%

Lieferant: Thermo Fisher Scientific
Achtung

437470010 437470050
ACRO437470010EA 159 EUR
ACRO437470010 ACRO437470050
(S)-3-(Aminomethyl)-1-Boc-piperidin 97%
(S)-3-(Aminomethyl)-1-Boc-piperidin
Formel: C₁₁H₂₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03839877
CAS-Nummer: 140645-24-5

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥96.0 %
Specific optical rotation -10° to -20° (20°C, 589 nm) (c=1, methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR