2-Methyl-2-propanyl-2-(hydroxymethyl)-1-piperidincarboxylat 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: tert-Butyl 2-(hydroxymethyl)piperidine-1-carboxylate, 1-Boc-2-hydroxymethylpiperidine, Tert-Butyl 2-(Hydroxymethyl)Tetrahydro-1(2H)-Pyridinecarboxylate, (±)-1-Boc-2-(hydroxymethyl)piperidine, BOC-2-piperidylmethanol

437520100
ACRO437520100EA 192 EUR
ACRO437520100
2-Methyl-2-propanyl-2-(hydroxymethyl)-1-piperidincarboxylat 97%
tert-Butyl-2-(hydroxymethyl)piperidine-1-carboxylate
Formel: C₁₁H₂₁NO₃
Molecular Weight: 215,293 g/mol
Schmelzpunkt: 74,4…75.7 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04114966
CAS-Nummer: 157634-00-9

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder or crystalline powder
Infrared spectrum Conforms
Melting point 73 to 77 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR