4-Methyl-1-naphthalinboronsäure 96%

Lieferant: Thermo Scientific
Achtung

437530100
ACRO437530100EA 207 EUR
ACRO437530100
4-Methyl-1-naphthalinboronsäure 96%
4-Methyl-1-naphthaleneboronic acid
Formel: C₁₁H₁₁BO₂
Molecular Weight: 186,02 g/mol
Schmelzpunkt: 224…228 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01632204
CAS-Nummer: 103986-53-4

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR