N-{[(2-Methyl-2-propanyl)oxy]carbonyl}-β-alanin 99%

Lieferant: Thermo Fisher Scientific

Synonyme: N-Boc-beta-alanine, Boc-β-Ala-OH, BOC-beta-alanine

437650050 437650250 437651000
ACRO437650050EA 20.4 EUR
ACRO437650050 ACRO437650250 ACRO437651000
N-{[(2-Methyl-2-propanyl)oxy]carbonyl}-β-alanin 99%
Boc-b-Alanine
Formel: C₈H₁₅NO₄
Schmelzpunkt: 77,5…78.5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00037291
CAS-Nummer: 3303-84-2

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder or solid
Infrared spectrum Conforms
Melting point 76 to 80 °C
Elemental Analysis 7.29 to 7.51 % N


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR