4-Cyanophenylacetic acid 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-(4-Cyanophenyl)acetic acid

437730010 437730050
ACRO437730010EA 117 EUR
ACRO437730010 ACRO437730050
4-Cyanophenylacetic acid 97%
4-Cyanophenylacetic acid
Formel: C₉H₇NO₂
Molecular Weight: 161.16 g/mol
Schmelzpunkt: 151…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06798066
CAS-Nummer: 5462-71-5
UN: 3439
ADR: 6.1 III

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 150.0 to 155.0 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR