4-Cyanophenylacetic acid 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-(4-Cyanophenyl)acetic acid
437730010
437730050
ACRO437730010EA
117
EUR
ACRO437730010
ACRO437730050
4-Cyanophenylacetic acid 97%
4-Cyanophenylacetic acid
Formel:
C₉H₇NO₂ Molecular Weight: 161.16 g/mol Schmelzpunkt: 151…152 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06798066 CAS-Nummer: 5462-71-5 UN: 3439 ADR: 6.1 III |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | 150.0 to 155.0 °C |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...