1-Benzyloxy-3-brombenzol 96%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
3-Benzyloxybromobenzene, 1-(Benzyloxy)-3-bromobenzene
437790100
437790500
ACRO437790100EA
52.8
EUR
ACRO437790100
ACRO437790500
1-Benzyloxy-3-brombenzol 96%
1-Benzyloxy-3-brombenzol
Formel:
C₁₃H₁₁BrO Molecular Weight: 263.13 Schmelzpunkt: 60…62 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00155065 CAS-Nummer: 53087-13-1 UN: 3077 ADR: 9 III |
Spezifikation Testergebnisse
Appearance | White Solid |
Infrared spectrum | Conforms |
Melting point | 59 to 64 °C |
GC | ≥95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...