4-Chlor-2,5-dimethoxyanilin 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2,5-Dimethoxy-4-chloroaniline
437861000
ACRO437861000EA
46.5
EUR
ACRO437861000
4-Chlor-2,5-dimethoxyanilin 97%
4-Chlor-2,5-dimethoxyanilin
Formel:
C₈H₁₀ClNO₂ Molecular Weight: 187,63 g/mol Schmelzpunkt: 118…120 °C Flammpunkt: 163 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014893 CAS-Nummer: 6358-64-1 |
Spezifikation Testergebnisse
Appearance | Off-white to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 118 to 120 °C |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...