2-Amino-3-brom-5-chlorpyridin 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-Brom-5-chlorpyridin-2-amin, 3-Brom-5-chlor-2-pyridylamin
437910010
437910050
ACRO437910010EA
45.8
EUR
ACRO437910010
ACRO437910050
2-Amino-3-brom-5-chlorpyridin 97%
2-Amino-3-brom-5-chlorpyridin
Formel:
C₅H₄BrClN₂ Molecular Weight: 207.46 g/mol Schmelzpunkt: 82…85 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00955627 CAS-Nummer: 26163-03-1 |
Spezifikation Testergebnisse
Appearance | White to light brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 83 to 87 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...