tert-Butyl-2-amino-6,7-dihydrothiazolo[5,4-c]pyridine-5(4H)-carboxylate 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
tert-Butyl 2-amino-6,7-dihydrothiazolo[5,4-c]pyridine-5(4H)-carboxylate, 5-BOC-2-AMINO-4,5,6,7-TETRAHYDROTHIAZOLO[5,4-C]PYRIDINE, 2-Amino-6,7-dihydro-4H-thiazolo[5,4-c]pyridine-5-carboxylicacidtert-butylester, tert-Butyl 2-amino-6,7-dihydro[1,3]thiazolo[5,4-c]pyridine-5(4H)-carboxylate
438020010
ACRO438020010EA
111
EUR
ACRO438020010
tert-Butyl-2-amino-6,7-dihydrothiazolo[5,4-c]pyridine-5(4H)-carboxylate 97%
tert-Butyl-2-amino-6,7-dihydrothiazolo[5,4-c]pyridine-5(4H)-carboxylate
Formel:
C₁₁H₁₇N₃O₂S Molecular Weight: 255,336 g/mol Schmelzpunkt: 223,5…224.5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08448157 CAS-Nummer: 365996-05-0 |
Spezifikation Testergebnisse
Appearance | Off-white to brown Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...