4-Benzyloxy-2-hydroxypyridin 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Benzyloxy-2(1H)-pyridone

438090050
ACRO438090050EA 166 EUR
ACRO438090050
4-Benzyloxy-2-hydroxypyridin 98%
4-Benzyloxy-2-hydroxypyridin
Formel: C₁₂H₁₁NO₂
Molecular Weight: 201,22 g/mol
Schmelzpunkt: 199,7…201.7 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00191783
CAS-Nummer: 53937-02-3

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to orange to brown Powder
Infrared spectrum Conforms
Melting point 199 to 203 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR