3,5-Di-tert-butylbenzoesäure 99%

Lieferant: Thermo Scientific
Achtung

438220050
ACRO438220050EA 106 EUR
ACRO438220050
3,5-Di-tert-butylbenzoesäure 99%
3,5-Di-tert-butylbenzoesäure
Formel: C₁₅H₂₂O₂
Molecular Weight: 234,34 g/mol
Schmelzpunkt: 170…173 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00082727
CAS-Nummer: 16225-26-6

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 170 to 175 °C
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR