2-Methyl-2-phenylpropanenitrile 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
438250010
438250050
ACRO438250010EA
68.6
EUR
ACRO438250010
ACRO438250050
2-Methyl-2-phenylpropanenitrile 97%
2-Methyl-2-phenylpropanenitrile
Formel:
C₁₀H₁₁N Siedepunkt: 54…56 °C (1 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00099614 CAS-Nummer: 1195-98-8 UN: 3276 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...