1,4-Dioxan-2,3-diol 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2,3-Dihydroxy-1,4-dioxane

438370050
ACRO438370050EA 93.9 EUR
ACRO438370050
1,4-Dioxan-2,3-diol 98%
1,4-Dioxane-2,3-diol
Formel: C₄H₈O₄
Molecular Weight: 120,10 g/mol
Schmelzpunkt: 92…94 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006572
CAS-Nummer: 4845-50-5

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Elemental Analysis 39.10 to 41.10 % (C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR