(R)-Methyl-2-((tert-butoxycarbonyl)amino)-3-iodopropanoate 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Boc-β-iodo-Ala-OMe, BOC-3-iodo-L-alanine methyl ester, N-Boc-3-iodo-L-alanine Methyl Ester, Boc-β-Iodo-Ala-OMe, Boc-Ala(3-I)-OMe

438460010 438460050
ACRO438460010EA 124 EUR
ACRO438460010 ACRO438460050
(R)-Methyl-2-((tert-butoxycarbonyl)amino)-3-iodopropanoate 97%
(R)-Methyl-2-((tert-butoxycarbonyl)amino)-3-iodopropanoate
Formel: C₉H₁₆INO₄
Schmelzpunkt: 49…52 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00216579
CAS-Nummer: 93267-04-0

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Powder
Infrared spectrum Conforms
Melting point 49 to 53 °C
HPLC ≥96.0 %
Specific optical rotation -3.0° to -5.0° (20 °C, 589 nm) (c=2 in methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR