tert-Butyl-3-(trifluoromethylsulphonyloxy)-8-azabicyclo[3.2.1]oct-3-ene-8-carboxylate 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 8-Azabicyclo[3.2.1]oct-2-ene-8-carboxylic acid,3-[[(trifluoromethyl)sulphonyl]oxy]-, 1,1-dimethylethyl ester, 3-(trifluoromethylsulphonyloxy)-8-azabicyclo[3.2.1]oct-3-ene-8-carboxylic acid tert-butyl ester, 8-Boc-3-(trifluoromethylsulphonyloxy)-8-azabicyclo[3.2.1]oct-3-ene

438590010
ACRO438590010EA 154 EUR
ACRO438590010
tert-Butyl-3-(trifluoromethylsulphonyloxy)-8-azabicyclo[3.2.1]oct-3-ene-8-carboxylate 95%
tert-Butyl-3-(trifluoromethylsulphonyloxy)-8-azabicyclo[3.2.1]oct-3-ene-8-carboxylate
Formel: C₁₃H₁₈F₃NO₅S
Molecular Weight: 357,344 g/mol
Schmelzpunkt: 36…40 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD14635483
CAS-Nummer: 185099-68-7

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder or low melting solid
Infrared spectrum Conforms
GC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR