3-(4-Methylphenyl)-1H-pyrazol-5-amin 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Amino-3-(4-methylphenyl)pyrazole, 3-Amino-5-(4-methylphenyl)-1H-pyrazole, 3-Amino-5-p-Tolylpyrazole, 5-(p-Tolyl)-1H-pyrazol-3-amine
438800050
ACRO438800050EA
174
EUR
ACRO438800050
3-(4-Methylphenyl)-1H-pyrazol-5-amin 97%
3-(4-Methylphenyl)-1H-pyrazol-5-amin
Formel:
C₁₀H₁₁N₃ Molecular Weight: 173,2 g/mol Schmelzpunkt: 152,3…153.3 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00084887 CAS-Nummer: 78597-54-3 |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | 150 to 154 °C |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...