O-Phenylhydroxylamine hydrochloride 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Hydroxylamine, O-phenyl-, hydrochloride, Phenoxyamine hydrochloride, N-phenoxyamine hydrochloride

438810010
ACRO438810010EA 156 EUR
ACRO438810010
O-Phenylhydroxylamine hydrochloride 97%
O-Phenylhydroxylamine hydrochloride
Formel: C₆H₈ClNO
Molecular Weight: 145,586 g/mol
Schmelzpunkt: 121…122 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00043271
CAS-Nummer: 6092-80-4
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white to light brown Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR