Adipic semialdehyde methyl ester 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
6-Oxohexanoic Acid Methyl Ester, Methyl 6-oxohexanoate, Hexanoic acid,6-oxo-,methyl ester
438912500
ACRO438912500EA
137
EUR
ACRO438912500
Adipic semialdehyde methyl ester 95%
Adipic semialdehyde methyl ester
Formel:
C₇H₁₂O₃ Molecular Weight: 144,17 g/mol Siedepunkt: 59…61 °C (1Torr) Lagertemperatur: Gefrierschrank |
MDL:
MFCD00063422 CAS-Nummer: 6654-36-0 |
Spezifikation Testergebnisse
Appearance | Colorless Liquid |
Infrared spectrum | Conforms |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...