(±)-Boc-α-phosphonoglycintrimethylester 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Methyl 2-((tert-butoxycarbonyl)amino)-2-(dimethoxyphosphoryl)acetate

439210010 439210050
ACRO439210010EA 45.6 EUR
ACRO439210010 ACRO439210050
(±)-Boc-α-phosphonoglycintrimethylester 97%
(±)-Boc-α-phosphonoglycintrimethylester
Formel: C₁₀H₂₀NO₇P
Schmelzpunkt: 69,7…71.6 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02178893
CAS-Nummer: 89524-98-1
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR