5-Bromgramin

Lieferant: Thermo Scientific
Gefahr

Synonyme: 1-(5-Bromo-1H-indol-3-yl)-N,N-dimethylmethanamine

439600010
ACRO439600010EA 67 EUR
ACRO439600010
5-Bromgramin
5-Bromgramin
Formel: C₁₁H₁₃BrN₂
Molecular Weight: 253,14 g/mol
Schmelzpunkt: 156…156,5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00055980
CAS-Nummer: 830-93-3
UN: 3259
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR