(3,4-Dimethoxybenzyl)methylamine 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 3,4-Dimethoxy-N-methylbenzylamine, N-Methyl-3,4-dimethoxybenzylamine

439680050
ACRO439680050EA 132 EUR
ACRO439680050
(3,4-Dimethoxybenzyl)methylamine 97%
(3,4-Dimethoxybenzyl)methylamine
Formel: C₁₀H₁₅NO₂
Molecular Weight: 181,24 g/mol
Flammpunkt: 110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00837593
CAS-Nummer: 63-64-9
UN: 2735
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Appearance Colorless to yellow to light brown Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR