2-Bromo-5-methoxybenzyl bromide 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
4-Bromo-3-(bromomethyl)anisole, 1-Bromo-2-(bromomethyl)-4-methoxybenzene
439790250
ACRO439790250EA
137
EUR
ACRO439790250
2-Bromo-5-methoxybenzyl bromide 97%
2-Bromo-5-methoxybenzyl bromide
Formel:
C₈H₈Br₂O Molecular Weight: 279,96 g/mol Schmelzpunkt: 91…93 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00078664 CAS-Nummer: 19614-12-1 UN: 3261 ADR: 8,II |
Spezifikation Testergebnisse
Appearance | White to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 90 to 94 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...