Monobenzyl malonate 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-(benzyloxy)-3-oxopropanoic acid, 2-((Benzyloxy)carbonyl)acetic acid, malonic acid monobenzyl ester

439800100
ACRO439800100EA 136 EUR
ACRO439800100
Monobenzyl malonate 95%
Monobenzyl malonate
Formel: C₁₀H₁₀O₄
Molecular Weight: 194,186 g/mol
Schmelzpunkt: 45…48 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00138132
CAS-Nummer: 40204-26-0

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 44 to 50 °C
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR