2-Brom-1-ethylpyridiniumtetrafluoroborat 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Bromo-1-ethyl-pyridinium tetrafluoroborate, 2-Bromo-1-ethylpyridin-1-ium tetrafluoroborate
439960050
ACRO439960050EA
88.9
EUR
ACRO439960050
2-Brom-1-ethylpyridiniumtetrafluoroborat 97%
2-Bromo-1-ethylpyridinium tetrafluoroborate
Formel:
C₇H₉BBrF₄N Molecular Weight: 273,86 g/mol Schmelzpunkt: 100…106 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00060046 CAS-Nummer: 878-23-9 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 100 to 108 °C |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...