3-Piperidin-1-ylpropanoic acid 96%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-Piperidinepropionic acid
440050250
ACRO440050250EA
82.2
EUR
ACRO440050250
3-Piperidin-1-ylpropanoic acid 96%
3-Piperidin-1-ylpropanoic acid
Formel:
C₈H₁₅NO₂ Molecular Weight: 157,21 g/mol Schmelzpunkt: 86…88,5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006515 CAS-Nummer: 26371-07-3 |
Spezifikation Testergebnisse
Appearance | White to light brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...