2-Fluorphenethylbromid 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
440120010
440120050
ACRO440120010EA
45.7
EUR
ACRO440120010
ACRO440120050
2-Fluorphenethylbromid 97%
2-Fluorphenethylbromid
Formel:
C₈H₈BrF Siedepunkt: 73…75 °C (20Torr) Lagertemperatur: Raumtemperatur |
MDL:
MFCD04117466 CAS-Nummer: 91319-54-9 UN: 3082 ADR: 9,III |
Spezifikation Testergebnisse
Appearance | Colorless to yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...